Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA6 Antibody (9J12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018052 0.025 mg
NB11093605 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018052 Supplier Novus Biologicals Supplier No. NB110936050.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphA6 Monoclonal specifically detects EphA6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EphA6
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9J12
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1000, Immunohistochemistry 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066
Gene Symbols EPHA6
Host Species Rat
Immunogen Recombinant mouse EPH Receptor A6 Antibody (9J12) extracellular domain
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285220
Test Specificity This was selected for its ability to detect mouse EPH Receptor A6 Antibody (9J12) in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.