Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA6 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB081969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081969 Supplier Novus Biologicals Supplier No. NBP298326F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EphA6 Polyclonal specifically detects EphA6 in Mouse samples. It is validated for ELISA.

Specifications

Antigen EphA6
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EphA6 (NP_031964.2; Met 1-Gln 546).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 285220
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.