Learn More
Description
Specifications
Specifications
| Antigen | EphA7 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 60 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 |
| Gene Symbols | EPHA7 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
