Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA7 Antibody (6E1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB110936060.025MG
Description
EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA7 | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1000, Immunohistochemistry-Paraffin | |
EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4. | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
60 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
EPHA7 | |
Recombinant mouse EphA7 extracellular domain | |
0.025 mg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
2045 | |
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction