Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA7 Antibody (6E1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | EphA7 |
---|---|
Clone | 60 |
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018053
![]() |
Novus Biologicals
NB110936060.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11093606
![]() |
Novus Biologicals
NB11093606 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA7 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
2045 | |
IgG1 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
60 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
EPHA7 | |
Primary | |
This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title