Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA7 Antibody (6E1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB193606549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB193606549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB193606549 Supplier Novus Biologicals Supplier No. NB11093606JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EphA7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 60
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3
Gene Symbols EPHA7
Host Species Rat
Immunogen Mouse EphA7 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2045
Test Specificity This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.