Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA7 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB081896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081896 Supplier Novus Biologicals Supplier No. NBP298316H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EphA7 Polyclonal specifically detects EphA7 in Mouse samples. It is validated for ELISA.

Specifications

Antigen EphA7
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EphA7 (Q61772-1; Met 1-Ile 556).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2045
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.