Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB1 Antibody (022), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB069164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069164 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069164 Supplier Novus Biologicals Supplier No. NBP290032AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EphB1 Monoclonal specifically detects EphB1 in Human samples. It is validated for ELISA.

Specifications

Antigen EphB1
Applications ELISA
Classification Monoclonal
Clone 22
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.1, EK6, ELK, EPH receptor B1, eph tyrosine kinase 2, EphB1, EPH-like kinase 6, ephrin type-B receptor 1, EPHT2FLJ37986, hEK6, NETHek6, soluble EPHB1 variant 1, Tyrosine-protein kinase receptor EPH-2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB1 (P54762-1; Met1-Pro540).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2047
Test Specificity No cross-reactivity in ELISA with: Human EPHB4 Human EPHB6 Mouse EphA1 Mouse EphA4 Mouse EphA6 Mouse EphA7 Mouse EphB3
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.