Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB2 Antibody (512012), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB369314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369314 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369314 Supplier Novus Biologicals Supplier No. FAB467M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphB2 Monoclonal specifically detects EphB2 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen EphB2
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 512012
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB2, Val27-Lys548 (predicted), Accession # P54763
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Prostate Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2048
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.