Learn More
Promega EPHB2-NanoLuc Fusion Vector, 20microg
Catalog No. PRNV1291 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRNV1291 20 μg
1 options
Catalog No. PRNV1291 Supplier Promega Supplier No. NV1291
Only null left
Add to Cart
Add to Cart
Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.
  • Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
  • Blue/white screening for easy identification of recombinant clones
  • In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • The difference between + and − vectors is orientation of the f1 origin
Specifications
Quantity 20 μg
Storage Requirements -30°C to -10°C