Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB3 Antibody (115), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB076022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076022 Supplier Novus Biologicals Supplier No. NBP290645AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EphB3 Monoclonal specifically detects EphB3 in Mouse samples. It is validated for ELISA.

Specifications

Antigen EphB3
Applications ELISA
Classification Monoclonal
Clone 115
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphB3 (NP_034273.1; Met 1-Thr 537).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2049
Test Specificity No cross-reactivity in ELISA with: Human LRPAP1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.