Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB4 Antibody (001), Biotin, Novus Biologicals™
SDP

Catalog No. NB066404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066404 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066404 Supplier Novus Biologicals Supplier No. NBP289352B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen EphB4
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 1
Conjugate Biotin
Dilution Flow Cytometry, ELISA
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB4 (NP_004435.3; Met1-Ala539).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2050
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.