Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB4 Antibody (395810), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313132 Supplier Novus Biologicals Supplier No. FAB3038APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EphB4
Applications Flow Cytometry
Classification Monoclonal
Clone 395810
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphB4, Ala16-Ala539, Accession # AAH52804
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2050
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.