Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB6 Antibody (031), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB066296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066296 Supplier Novus Biologicals Supplier No. NBP289327R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen EphB6
Applications ELISA
Classification Monoclonal
Clone 31
Conjugate DyLight 550
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 (NP_004436.1; Met1-Ser579).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2051
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.