Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB6 Antibody (465327), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312957 Supplier Novus Biologicals Supplier No. FAB33841PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EphB6
Applications Flow Cytometry
Classification Monoclonal
Clone 465327
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphB6, Val17-Thr579, Accession # O15197
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2051
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.