Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A1 Antibody (114), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB067683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067683 Supplier Novus Biologicals Supplier No. NBP289691IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Ephrin-A1
Applications ELISA
Classification Monoclonal
Clone 114
Conjugate DyLight 755
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Ephrin-A1 (NP_004419.2; Met 1-Ser 182).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1942
Test Specificity No cross-reactivity in ELISA with: Human EphrinA5/ EFNA5 Human EphrinA3/ EFNA3
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.