Learn More
Description
Specifications
Specifications
| Antigen | Ephrin-A1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 114 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Ephrin-A1 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
