Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB081256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081256 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081256 Supplier Novus Biologicals Supplier No. NBP298103V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ephrin-A2 Polyclonal specifically detects Ephrin-A2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Ephrin-A2
Applications ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Ephrin-A2 (NP_031935.3; Met 1-Asn 184).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1943
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.