Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A3 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB053362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053362 Supplier Novus Biologicals Supplier No. NBP298317V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ephrin-A3 Polyclonal specifically detects Ephrin-A3 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ephrin-A3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Ephrin-A3 (NP_034238.1; Met1-Ser205).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1944
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.