Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A3 Antibody (MM0262-7N12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP212279B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212279B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212279B Supplier Novus Biologicals Supplier No. NBP212279B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ephrin-A3 Monoclonal specifically detects Ephrin-A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ephrin-A3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0262-7N12
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3
Gene Symbols EFNA3
Host Species Mouse
Immunogen Human recombinant Ephrin-A3 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1944
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.