Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A4 Antibody (MM0263-9H13), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP212280A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP212280A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP212280A32 Supplier Novus Biologicals Supplier No. NBP212280AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ephrin-A4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0263-9H13
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
Gene Symbols EFNA4
Host Species Mouse
Immunogen Human recombinant Ephrin-A4 (extracellular domain)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1945
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.