Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-B3 Antibody (MM0264- 12X1), PerCP, Novus Biologicals™
SDP

Catalog No. NB361108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361108 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361108 Supplier Novus Biologicals Supplier No. NBP212283PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ephrin-B3 Monoclonal antibody specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Ephrin-B3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0264- 12X1
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6
Host Species Mouse
Immunogen Human recombinant Ephrin-B3 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.