Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHX1 Antibody (2E5), HRP, Novus Biologicals™
SDP

Catalog No. NBP130163H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130163H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130163H Supplier Novus Biologicals Supplier No. NBP130163H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EPHX1 Monoclonal specifically detects EPHX1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen EPHX1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 200000
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Gene Alias EC 3.3.2.9, EPHX, EPOX, Epoxide hydratase, epoxide hydrolase 1, epoxide hydrolase 1, microsomal (xenobiotic), MEH, Microsomal epoxide hydrolase
Gene Symbols EPHX1
Host Species Mouse
Immunogen Recombinant human EPHX1 (21-455aa) purified from E. coli
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2052
Test Specificity This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human EPHX1 protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.