Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunoprecipitation experiments with Epredia™ Epidermal Growth Factor Receptor/EGFR Ab-2, Mouse Monoclonal Antibody.

$298.00 - $593.00

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-2
Clone 225
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS269P0
View Documents
Epredia
MS269P0
200μg/mL 100 μL
Each for $298.00
Only null left
Add to Cart
 
MS269P1
View Documents
Epredia
MS269P1
200μg/mL 500 μL
Each for $593.00
Only null left
Add to Cart
 
MS269P
View Documents
Epredia
MS269P
200μg/mL 1 mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS269P1ABX
View Documents
Epredia
MS269P1ABX
1mg/mL 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS269PABX
View Documents
Epredia
MS269PABX
1mg/mL 200 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 225

Isotype: IgG1

Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native only)
  • Blocks EGF/TGFα-Induced Activation of EGFR
  • Arrests Tumor Growth in vivo
  • Inhibits Binding of Ab-1 (clone 528) to EGFR

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epidermal Growth Factor Receptor/EGFR Ab-2
Immunoprecipitation
Unconjugated
Mouse
EGFR Family
Purified EGFR from A431 cells
Primary
225
Monoclonal
Concentrated
RUO
Human, Non-human Primate
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.