Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epimorphin/Syntaxin 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018391 0.025 mg
NBP119338 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018391 Supplier Novus Biologicals Supplier No. NBP1193380.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Epimorphin/Syntaxin 2 Polyclonal specifically detects Epimorphin/Syntaxin 2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Epimorphin/Syntaxin 2
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml (colorimetric), Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias EPIMepimorphin, EPM, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, syntaxin 2
Gene Symbols STX2
Host Species Rabbit
Immunogen Synthetic peptide derived from sequence near the amino-terminus of rat Syntaxin 2, conjugated to KLH
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2054
Test Specificity Syntaxin 2
Target Species Xenopus species, Chicken, Pig, Mouse, Drosophila, Monkey, Rat, Rabbit, Bovine, Hamster, Canine, Human, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.