Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epimorphin/Syntaxin 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $560.50

Specifications

Antigen Epimorphin/Syntaxin 2
Concentration 1.0 mg/mL
Dilution Western Blot 1 ug/ml (colorimetric), Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018391
SDP
View Documents
Novus Biologicals
NBP1193380.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP119338
SDP
View Documents
Novus Biologicals
NBP119338
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Epimorphin/Syntaxin 2 Polyclonal specifically detects Epimorphin/Syntaxin 2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Epimorphin/Syntaxin 2
Western Blot 1 ug/ml (colorimetric), Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Polyclonal
Rabbit
Xenopus species, Chicken, Pig, Mouse, Drosophila, Monkey, Rat, Rabbit, Bovine, Hamster, Canine, Human, Sheep
EPIMepimorphin, EPM, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, syntaxin 2
STX2
IgG
Affinity Purified
Syntaxin 2
1.0 mg/mL
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
2054
Synthetic peptide derived from sequence near the amino-terminus of rat Syntaxin 2, conjugated to KLH
Primary
Store at -20C. Avoid freeze-thaw cycles.
35 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.