Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epiregulin Antibody (183629), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB297137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297137 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297137 Supplier Novus Biologicals Supplier No. FAB14251G

Mouse Monoclonal Antibody

Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Epiregulin
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 183629
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias epiregulin, ER, proepiregulin
Host Species Mouse
Immunogen E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2069
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.