Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epiregulin Antibody (183629) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB400032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400032 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400032 Supplier Novus Biologicals Supplier No. FAB14251I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen Epiregulin
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 183629
Conjugate Janelia Fluor 549
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias epiregulin, ER, proepiregulin
Host Species Mouse
Immunogen E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2069
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.