Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epiregulin Antibody (6H12), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB360843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360843 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360843 Supplier Novus Biologicals Supplier No. NBP221992MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Epiregulin Monoclonal antibody specifically detects Epiregulin in Mouse samples. It is validated for ELISA, Neutralization

Specifications

Antigen Epiregulin
Applications ELISA, Neutralization
Classification Monoclonal
Clone 6H12
Conjugate mFluor Violet 450 SE
Dilution ELISA, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias epiregulin, ER, proepiregulin
Host Species Rat
Immunogen Mouse recombinant Epiregulin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 2069
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.