Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelia (colonic) Antibody (PR 4D1), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310385 Supplier Novus Biologicals Supplier No. NB1002689MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Epithelia (colonic) Monoclonal specifically detects Epithelia (colonic) in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Epithelia (colonic)
Applications Immunohistochemistry
Classification Monoclonal
Clone PR 4D1
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Mucosal scrapings and membrane preparations from normal colorectal epithelium and possibly booster inoculations with the colon carcinoma cell line HT29.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.