Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial cell marker Antibody (LH39), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310384 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310384 Supplier Novus Biologicals Supplier No. NB1002690MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Epithelial cell marker Monoclonal specifically detects Epithelial cell marker in Primate samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Epithelial cell marker
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LH39
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.