Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epithelial Sodium Channel alpha Antibody (2G4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP259382IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259382IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259382IR Supplier Novus Biologicals Supplier No. NBP259382IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

epithelial Sodium Channel alpha Monoclonal specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen epithelial Sodium Channel beta
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2G4
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Gene Symbols SCNN1B
Host Species Mouse
Immunogen Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68)
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6338
Test Specificity Detects 85kDa.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.