Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel alpha Antibody (2G4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259382
Description
epithelial Sodium Channel alpha Monoclonal specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
epithelial Sodium Channel beta | |
Monoclonal | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
SCNN1B | |
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68) | |
100 μg | |
Phospho Specific | |
6338 | |
Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
2G4 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects 85kDa. | |
Store at -20C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction