Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel alpha Antibody (2G4), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | epithelial Sodium Channel beta |
---|---|
Clone | 2G4 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
epithelial Sodium Channel alpha Monoclonal specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
epithelial Sodium Channel beta | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
SCNN1B | |
IgG2b | |
Protein G purified |
2G4 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Phospho Specific | |
6338 | |
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68) | |
Primary | |
Detects 85kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title