Learn More
epithelial Sodium Channel beta Antibody (7B8), DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | epithelial Sodium Channel beta |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 7B8 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta |
| Gene Symbols | SCNN1B |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.