Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epithelial Sodium Channel gamma Antibody (3c7), Biotin, Novus Biologicals™
SDP

Catalog No. NBP241371B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241371B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP241371B Supplier Novus Biologicals Supplier No. NBP241371B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

epithelial Sodium Channel gamma Monoclonal specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA.

Specifications

Antigen epithelial Sodium Channel gamma
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3c7
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma
Gene Symbols SCNN1G
Host Species Mouse
Immunogen The inhibitory peptide from the human gamma-ENaC subunit. EAESWNSVSEGKQPRFSHRIPLC corresponding to amino acid residue 139-160 of human gamma-ENaC subunit.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6340
Test Specificity Antibody is specific for the inhibitory tract of human gamma-ENaC subunit.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.