Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epithelial Sodium Channel gamma Mouse anti-Human, Clone: 5c2, Novus Biologicals™
SDP

Catalog No. NB019682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019682 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019682 Supplier Novus Biologicals Supplier No. NBP2413730.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

epithelial Sodium Channel gamma Monoclonal specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA.

Specifications

Antigen epithelial Sodium Channel gamma
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone 5c2
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Sandwich ELISA
Formulation 0.01 M phosphate buffer, pH 7.4, containing 0.5 M NaCl
Gene Alias amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma
Host Species Mouse
Immunogen The inhibitory peptide from the human gENaC subunit. EAESWNSVSEGKQPRFSHRIPLC corresponding to amino acid residue 139-160 of human gENaC subunit.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6340
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.