Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPLIN Antibody (OTI1A3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271932A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271932A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271932A700 Supplier Novus Biologicals Supplier No. NBP271932AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EPLIN Monoclonal specifically detects EPLIN in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EPLIN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A3
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3
Gene Symbols LIMA1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-275 of human EPLIN (NP_057441) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51474.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.