Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPM2AIP1 Antibody (OTI2A2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271497G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271497G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271497G Supplier Novus Biologicals Supplier No. NBP271497G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EPM2AIP1 Monoclonal specifically detects EPM2AIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen EPM2AIP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2A2
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein
Gene Symbols EPM2AIP1
Host Species Mouse
Immunogen Full length recombinant protein of human EPM2AIP1 (NP_055620) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9852.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.