Learn More
Invitrogen™ EpoR Recombinant Rabbit Monoclonal Antibody (4O8U2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557949

Description
Immunogen sequence: APPPNLPDPK FESKAALLAA RGPEELLCFT ERLEDLVCFW EEAASAGVGP GNYSFSYQLE DEPWKLCRLH QAPTARGAVR FWCSLPTADT SSFVPLELRV TAASGAPRYH RVIHINEVVL LDAPVGLVAR LADESGHVVL RWLPPPETPM TSHIRYEVDV SAGNGAGSVQ RVEILEGRTE CVLSNLRGRT RYTFAVRARM AEPSFGGFWS AWSEPVSLLT PSDLD.
EpoR encodes the erythropoietin receptor, which belongs to the cytokine receptor family. When erythropoietin binds to this receptor, it activates the Jak2 tyrosine kinase, which in turn activates various intracellular pathways including Ras/MAP kinase, phosphatidylinositol 3-kinase, and STAT transcription factors. The stimulated erythropoietin receptor plays a role in the survival of erythroid cells. Defects in this gene may result in erythroleukemia and familial erythrocytosis. Dysregulation of this gene may also affect the growth of certain tumors. Multiple transcript variants are produced as a result of alternate splicing. Diseases associated with EPOR include Erythrocytosis, Familial, 1 and Erythroleukemia.
Specifications
EpoR | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P19235 | |
EPOR | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-249 of human EPOR (NP_000112.1). | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
4O8U2 | |
Unconjugated | |
EPOR | |
EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR | |
Rabbit | |
Protein A | |
RUO | |
2057 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.