Learn More
Eppendorf™ 5804/5810 Series Horizontal Drum Rotor Tube Adapters
Catalog No. p-29176 Shop All Eppendorf North America Products
Change view
Click to view available options
Tube Size:
1.5 to 2 mL
Catalog No. Tube Size
05-40-102 1.5 to 2 mL
1 options
This item is not returnable. View return policy
Catalog No. 05-40-102 Supplier Eppendorf™ Supplier No. 022638521
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with rotor model T-60-11
Adapters are supplied in packs of six. Order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor for 5804/5804R/5810/5810R centrifuge
Tube Size 1.5 to 2 mL