Learn More
Eppendorf™ Combitips™ Tips
Catalog No. p-161400 Shop All Eppendorf North America Products
Click to view available options
Volume (Metric):
12.5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
For Eppendorf Repeater™ Pipetter
Tips have polyethylene plunger and polypropylene cylinder, making them ideal for use with most reagents
Order Info
Nonsterile tips are packed in plastic bags. Biopur™ tips are individually wrapped.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 12.5 mL
Sterility Sterile
Tip Style Specialty
For Use With (Equipment) Eppendorf Repeater
Product Line CombiTips
Material Polypropylene Cylinder, Polyethylene Plunger
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.