Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Femtotips™ Microloader Tips for Femtojet Microinjector

Catalog No. e5242956003 Shop All Eppendorf North America Products
Click to view available options
For Use With (Equipment):
Femtojet microinjector

Catalog No. E5242956003

Supplier: Eppendorf™ 930001007

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Tips to fill microinjection capillaries and recovering surplus solution from capillary

  • With Stringent quality-control
  • Non-filter tip
  • Universal tip fit
  • Volume range: 0.5 - 20μL
  • Color: Light gray
  • Sterilizable rack package

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For recovering surplus solution from the capillary
For Use With (Equipment) Femtojet microinjector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.