Eppendorf™ PCR Clean Microcentrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.

Choose from Safe-Lock or Flex-Tube™ styles—both are ideal for DNA sample prep prior to PCR

Manufacturer: Eppendorf™ 022363212

 View more versions of this product

Catalog No. 05-402-94

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 1.5mL
    Color Clear
    Material Polypropylene
    Max. Force 30000xG
    Sterile No
    For Use With (Application) Ideal for DNA sample prep prior to PCR
    Graduated Yes
    Certifications/Compliance Human DNA, DNase, RNase, PCR inhibitor-free
    Shape Conical
    Autoclavable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit