Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Tubes™ with Snap Cap 5.0 mL - Microtube
GREENER_CHOICE

Catalog No. 13864404 Shop All Eppendorf North America Products
Change view
Click to view available options
Packaging:
10 Bags x 20 Tubes
2 Bags x 100 Tubes
4 Bags x 100 Tubes
4 Bags x 50 Tubes
Individually Wrapped
Purity or Quality Grade:
Biopur™
Eppendorf Quality
Forensic DNA Grade
PCR Clean
Sterile
Color:
Amber
Clear
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Purity or Quality Grade Color
13-864-404 2 Bags x 100 Tubes PCR Clean Amber
14-282-300 2 Bags x 100 Tubes Eppendorf Quality Clear
14-282-301 2 Bags x 100 Tubes PCR Clean Clear
14-282-306 4 Bags x 100 Tubes PCR Clean Clear
E0030119606 4 Bags x 50 Tubes Forensic DNA Grade Clear
14-282-302 10 Bags x 20 Tubes Sterile Clear
14-282-303 Individually Wrapped Biopur™ Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 13-864-404 Supplier Eppendorf™ Supplier No. 0030119452
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eppendorf Tubes™ with snap caps are your ideal choice when working with medium-sized sample volumes (0.5 mL to 5.0 mL). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25,000 x g), and sample recovery.

Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.

  • Ideal for working with medium-sized sample volumes ranging from 0.5 mL to 5.0 mL.
  • Designed for ease of use and user-friendly handling during sample preparation and processing.
  • Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions.
  • Provides reliable safety during centrifugation with a maximum rating of up to 25,000 x g.
  • Specifically designed to enhance sample recovery, preserving the integrity of your samples.
  • Available in Eppendorf Quality, PCR Clean, Biopur™, Sterile, and Forensic DNA Grade quality for tested purity.
  • Free of plasticizers, biocides or mold release agents.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Eppendorf Quality certified purity
Closure Material Polypropylene
Color Amber
Capacity (Metric) 5 mL
Material Polypropylene
For Use With Processing samples up to 5.0mL; DNA isolation, especially with samples of limited quantity and source material; Preparation of mastermixes and buffers; Cell and tissue lysis
No. per Pack 200 Pk.
Packaging 2 Bags x 100 Tubes
Quantity 200 Tubes
Sterility Non-sterile
Autoclavable Autoclavable
Shape Cylindrical
Bottom Shape Conical
Closure Type Snap Cap
Diameter (Metric) 17 mm
Graduated Yes
Max. RCF 25,000 x g
Purity or Quality Grade PCR Clean
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.