Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Shop All Eppendorf North America Products

Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Skirt Style:
Skirted
Unskirted
Unskirted, Divisible
Product Type:
Divisible PCR Plates
Forensic DNA Grade PCR Plates
Low Profile PCR Plates
Microbiology PCR Plates
PCR Plates
Standard PCR Plate
Packaging:
10 Bags x 30 Plates
5 Bags x 4 Plates
5 Bags x 5 Plates
Individually Blister Wrapped
Description
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- Improved well-to-well tolerance.
- Cut-away corner and alphanumeric grid-referencing.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
† Certificate, test procedures and detailed information available upon request.
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | Human DNA, DNase, RNase-free |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | Individually Blister Wrapped |
| Color | Blue |
| Product Type | Microbiology PCR Plates |
| Skirt Style | Skirted |
| Material | Polypropylene, Polycarbonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction