Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.5

Catalog No. p-7042145
Click to view available options
Quantity:
100 mL
250 mL

Catalog No. AAJ61476AE

Supplier: Thermo Scientific Chemicals J61476AE

Only null left
Add to Cart

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material EPPS buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.