Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPRS Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290264 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290264 Supplier Novus Biologicals Supplier No. NBP222311PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPRS Polyclonal antibody specifically detects EPRS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen EPRS
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias bifunctional aminoacyl-tRNA synthetase, Cell proliferation-inducing gene 32 protein, DKFZp313B047, EARS, EC 6.1.1, EC 6.1.1.15, EC 6.1.1.18, GLNS, GLUPRORS, glutamate tRNA ligase, glutaminyl-tRNA synthetase, glutamyl-prolyl-tRNA synthetase, PARSproliferation-inducing gene 32 protein, proliferation-inducing protein 32, proline tRNA ligase, proline-tRNA ligase, prolyl-tRNA synthetase, QARS, QPRS
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 950 to 1000 of human Glutamyl-prolyl-tRNA Synthetase using the numbering given in entry NP_004437.2 (GeneID 2058).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2058
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.