Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS15 Antibody (1261C), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB302555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302555 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302555 Supplier Novus Biologicals Supplier No. FAB8480N

Rabbit Monoclonal Antibody

EPS15 Monoclonal antibody specifically detects EPS15 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen EPS15
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1261C
Conjugate Alexa Fluor 700
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15
Host Species Rabbit
Immunogen E.coli-derived recombinant human Eps15, Ala448-Thr579, Accession # P42566
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2060
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.