Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS15R Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095599 100 μL
NB095600 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095599 Supplier Novus Biologicals Supplier No. NBP303739100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPS15R Polyclonal specifically detects EPS15R in Human samples. It is validated for Western Blot.

Specifications

Antigen EPS15R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias epidermal growth factor receptor pathway substrate 15-like 1, epidermal growth factor receptor substrate 15-like 1, epidermal growth factor receptor substrate EPS15R, EPS15R, Eps15-related protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 475-754 of human EPS15R (NP_001245304.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58513
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.